Learn More
Medchemexpress LLC HY-N0496 5mg Medchemexpress, Ruscogenin CAS:472-11-7 Purity:>98%
SDP
Catalog No. 501933596
Encompass_Preferred
Medchemexpress, HY-N0496 5mg Ruscogenin CAS:472-11-7 Ruscogenin, an important steroid sapogenin derived from Ophiopogon japonicus, attenuates cerebral ischemia-induced blood-brain barrier dysfunction by suppressing TXNIP/NLRP3 inflammasome activation and the MAPK pathway and exerts significant anti-inflammatory and anti-thrombotic activities [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3596 Supplier Medchemexpress LLC Supplier No. HYN04965MG
Medchemexpress, HY-N0496 5mg Ruscogenin CAS:472-11-7 Ruscogenin, an important steroid sapogenin derived from Ophiopogon japonicus, attenuates cerebral ischemia-induced blood-brain barrier dysfunction by suppressing TXNIP/NLRP3 inflammasome activation and the MAPK pathway and exerts significant anti-inflammatory and anti-thrombotic activities [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.