Learn More
MSC RUSH/ PU SAFETY GLASSES
Catalog No. 5000687991
Encompass
Bollé Safety RUSH+ safety glasses feature an ultra-sporty, half-framed, wrap-around design with co-injected and ultra-flexible temples. They are equipped with Platinum anti-fog and anti-scratch clear polycarbonate lenses.
  • Bi-material, ultra-flexible colored temples
  • Non-slip adjustable bridge
  • Upper protection
  • UV protection
  • Meets ANSI Z87.1-2010 standards
  • Supplied with microfiber pouch
Catalog No. 50-006-87991 Supplier MSC Supplier No. 85776730
Bollé Safety RUSH+ safety glasses feature an ultra-sporty, half-framed, wrap-around design with co-injected and ultra-flexible temples. They are equipped with Platinum anti-fog and anti-scratch clear polycarbonate lenses.
  • Bi-material, ultra-flexible colored temples
  • Non-slip adjustable bridge
  • Upper protection
  • UV protection
  • Meets ANSI Z87.1-2010 standards
  • Supplied with microfiber pouch