Learn More
Chemglass Life Sciences Rushton Style Impeller Blade for 2-3L Bio Reactors, 46mm Dia for 8mm OD Shaft
SDP
Catalog No. NC3239085
Encompass
This adapter can be used to add fluids or gases to the bio reactor.The portion of the adapter that is within the reactor has a short drip tip tube that can accept tubing to reach further into the listed capacity reactor.Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc.Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. NC3239085 Supplier Chemglass Life Sciences Supplier No. CLS138008
This adapter can be used to add fluids or gases to the bio reactor.The portion of the adapter that is within the reactor has a short drip tip tube that can accept tubing to reach further into the listed capacity reactor.Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc.Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.