Learn More
Chemglass Life Sciences Rushton Style Impeller Blade for 15L Bio Reactors, 76mm Dia for 8mm OD Shaft
SDP
Catalog No. 501294562
Encompass
Rushton Style Impeller Blade for 15L Bio Reactors, 76mm Dia, 8mm Shaft Size. The 3 through 7 Liter sizes fit the 8mm Shaft, while the 15 liter fits the 15mm shaft. These blades are used primarily for bacterial cell culture work.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4562 Supplier Chemglass Life Sciences Supplier No. CLS138550
Rushton Style Impeller Blade for 15L Bio Reactors, 76mm Dia, 8mm Shaft Size. The 3 through 7 Liter sizes fit the 8mm Shaft, while the 15 liter fits the 15mm shaft. These blades are used primarily for bacterial cell culture work.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.