Learn More
Bellco Glass, Inc. Rushton Impeller Assembly ,15L
SDP
Catalog No. 501946480
Encompass_Preferred
Rushton impeller 2”OD with 6 vertical blades equally spaced around the center hub. Manufactured from stainless steel and designed for high speed fermentation protocols. Assembled onto 3L metric bioreactor drive stem.. MADE IN THE USA
Catalog No. 50-194-6480 Supplier Bellco Glass, Inc. Supplier No. 790915054
Rushton impeller 2”OD with 6 vertical blades equally spaced around the center hub. Manufactured from stainless steel and designed for high speed fermentation protocols. Assembled onto 3L metric bioreactor drive stem.. MADE IN THE USA