Learn More
Bellco Glass, Inc. Rushton Impeller Assembly ,7L
SDP
Catalog No. 501946447
Encompass_Preferred
Rushton impeller 2”OD with 6 vertical blades equally spaced around the center hub. Manufactured from stainless steel and designed for high speed fermentation protocols. Assembled onto 3L metric bioreactor drive stem.. MADE IN THE USA
Catalog No. 50-194-6447 Supplier Bellco Glass, Inc. Supplier No. 790900040
Rushton impeller 2”OD with 6 vertical blades equally spaced around the center hub. Manufactured from stainless steel and designed for high speed fermentation protocols. Assembled onto 3L metric bioreactor drive stem.. MADE IN THE USA