Learn More
Stoelting™ Ruskin Bone Cutter
SDP
Catalog No. 10001806 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Straight
Catalog No. Model
10-001-806 Straight
1 options
Catalog No. 10-001-806 Supplier Stoelting™ Supplier No. 5216775
Add to Cart
Add to Cart
Bone cutting tools allow for sharp cuts through soft bone and cartilage.
Specifications
For Use With (Application) Neuroscience
Length (English) 5.5 in.
Model Straight