Learn More
Electron Microscopy Sciences Russell's Modified Zenkers Solution 500 ML
SDP
Catalog No. 5032085
Encompass_Preferred
Due to the hazardous nature of mercury, Zenker's fluid is no longer available. In place we are offering Russell's Zenker Fixative, which contains Zinc Chloride. The staining results produced by this fixative are comparable to those of Zenker's Stock solution. Russell's Modified Zenker's is used in the same manner as Zenker's stock solution.
Catalog No. 50-320-85 Supplier Electron Microscopy Sciences Supplier No. 6412305
Due to the hazardous nature of mercury, Zenker's fluid is no longer available. In place we are offering Russell's Zenker Fixative, which contains Zinc Chloride. The staining results produced by this fixative are comparable to those of Zenker's Stock solution. Russell's Modified Zenker's is used in the same manner as Zenker's stock solution.