Learn More
Electron Microscopy Sciences Russels Modified Zenkers Fluid 500 ML
SDP
Catalog No. 5031879
Encompass_Preferred
Used in Ptah Method for Central Nervous Tissue for staining microglia, fibroglia, fibrin and coarse fibrils. References: AFIP Manual of Histologic Staining Techniques; (ed) G. Luna, 3rd Ed. McGraw-Hill Publications, c. 1968, p. 199
Catalog No. 50-318-79 Supplier Electron Microscopy Sciences Supplier No. 2665805
Used in Ptah Method for Central Nervous Tissue for staining microglia, fibroglia, fibrin and coarse fibrils. References: AFIP Manual of Histologic Staining Techniques; (ed) G. Luna, 3rd Ed. McGraw-Hill Publications, c. 1968, p. 199