Learn More
Electron Microscopy Sciences Russian Tissue Forceps 8"
SDP
Catalog No. 5028409
Encompass_Preferred
Russian tissue forceps. Stainless steel 8" long, with special cupped pattern for superior grip, cup area is 6.5mm wide, 9mm long.
Catalog No. 50-284-09 Supplier Electron Microscopy Sciences Supplier No. 7826008
Russian tissue forceps. Stainless steel 8" long, with special cupped pattern for superior grip, cup area is 6.5mm wide, 9mm long.