Learn More
Thermo Scientific Rust Inhibitor
Catalog No. NC1828108 Shop All Thermo Scientific Products
Encompass_Preferred
Used with any Symphony Water Jacket Incubator. Rust Inhibitor should be used anytime the water jacket is emptied or every two years.
Catalog No. NC1828108 Supplier Thermo Scientific Supplier No. 1900101
Used with any Symphony Water Jacket Incubator. Rust Inhibitor should be used anytime the water jacket is emptied or every two years.