Learn More
Enterprise Technology Solutions Rust-Oleum Ceiling Paint/Primer, Vertical Spray, 13 oz, WE
SDP
Catalog No. 5000829087
Encompass_Preferred
Ceiling paint and primer spray features a vertical aerosol spray that delivers a smooth, even blast straight up to efficiently apply directly to ceilings. Advanced, low-odor formula offers a quick cure for water stains to easily extend the life of stained or dingy ceilings. The flat white finish is nonyellowing and matches most ceiling tiles to blend flawlessly with your ceiling.
Catalog No. 50-008-29087 Supplier Enterprise Technology Solutions Supplier No. RST3688
Add to Cart
Add to Cart
Ceiling paint and primer spray features a vertical aerosol spray that delivers a smooth, even blast straight up to efficiently apply directly to ceilings. Advanced, low-odor formula offers a quick cure for water stains to easily extend the life of stained or dingy ceilings. The flat white finish is nonyellowing and matches most ceiling tiles to blend flawlessly with your ceiling.