Learn More
Enterprise Technology Solutions Rust-Oleum Coating,AbrasionResistant,UrethaneLatexEpoxy,White
SDP
Catalog No. 5000790688
Encompass_Preferred
The MULE (Modified Urethane Latex Epoxy) delivers ultra-smooth finishes to both indoor and outdoor surfaces. Abrasion-resistant coating is perfect for high-traffic areas subject to routine cleaning and disinfecting. Low-odor, highly washable formula easily adheres to prepared and primed surfaces including metal, concrete, brick, cinderblock, wood and drywall. Use in commercial spaces, offices, schools, health care facilities and more.
Catalog No. 50-007-90688 Supplier Enterprise Technology Solutions Supplier No. RST375658
The MULE (Modified Urethane Latex Epoxy) delivers ultra-smooth finishes to both indoor and outdoor surfaces. Abrasion-resistant coating is perfect for high-traffic areas subject to routine cleaning and disinfecting. Low-odor, highly washable formula easily adheres to prepared and primed surfaces including metal, concrete, brick, cinderblock, wood and drywall. Use in commercial spaces, offices, schools, health care facilities and more.