Learn More
Enterprise Technology Solutions Rust-Oleum Degreaser,f/Nonporous,Concentrated,Bottle,1 Gallon,4/CT,CL
SDP
Supplier:  Enterprise Technology Solutions RST352261CT
Encompass_Preferred
Quickly and effectively remove tough soils from many different surfaces using this Concentrated Cleaner Degreaser. Heavy-duty formula dissolves grease and grime on most nonporous surfaces. Degreaser is perfect for industrial cleaning without the use of harsh chemicals. Fabric-safe formula leaves no residue behind. Use this degreaser to remove grime, oil, tar, wax and more from items such as tools, machinery, engine parts, construction equipment, fiberglass, nonfinished concrete, masonry and asphalt. The 1 gallon supply rids surfaces of larger stains with ease.
Catalog No. 50-007-90956