Learn More
Enterprise Technology Solutions Rust-Oleum Graffiti Remover, Water-Based, Multi-surface, 32 fl. oz, MI
SDP
Catalog No. 5000790684
Encompass_Preferred
Graffiti remover features a nonflammable, water-based formula to provide a safe alternative to hazardous, solvent-based graffiti removers. Highly versatile design is safe for use on most hard, soft and porous surfaces, including aluminum, concrete, freeway signs, plastic, split rock, tile, brick, glass, street signs, vinyl and more. Plus, it's safe for use on clear coat automotive finishes. Graffiti remover is perfect for removing acrylic, enamel, primer, sealers, stain killers, aerosol paint, oil lacquers and more.
Catalog No. 50-007-90684 Supplier Enterprise Technology Solutions Supplier No. RSTGR326
Graffiti remover features a nonflammable, water-based formula to provide a safe alternative to hazardous, solvent-based graffiti removers. Highly versatile design is safe for use on most hard, soft and porous surfaces, including aluminum, concrete, freeway signs, plastic, split rock, tile, brick, glass, street signs, vinyl and more. Plus, it's safe for use on clear coat automotive finishes. Graffiti remover is perfect for removing acrylic, enamel, primer, sealers, stain killers, aerosol paint, oil lacquers and more.