Learn More
Enterprise Technology Solutions Rust-Oleum Marking Paint, 17 oz., Water Based, Fluor Orange
SDP
Catalog No. 5000829610
Encompass_Preferred
Create ultra-crisp, long-lasting marks with Industrial Choice M1800 System Precision Line Inverted Marking Paint Spray. This water-based formula creates accurate, easy-to-read markings in a bright, durable color. Use it for marking concrete, pavement, gravel, grass and more. Paint spray is ideal for marking the location of underground utilities at construction and excavation sites where identification is desired. The precision tip means virtually no overspray. Paint resists early fading and chalking. It adheres to hot surfaces and can be sprayed in the cold (to 25 degrees Fahrenheit). Once applied, the paint dries in under 5 minutes.
Catalog No. 50-008-29610 Supplier Enterprise Technology Solutions Supplier No. RST203036
Create ultra-crisp, long-lasting marks with Industrial Choice M1800 System Precision Line Inverted Marking Paint Spray. This water-based formula creates accurate, easy-to-read markings in a bright, durable color. Use it for marking concrete, pavement, gravel, grass and more. Paint spray is ideal for marking the location of underground utilities at construction and excavation sites where identification is desired. The precision tip means virtually no overspray. Paint resists early fading and chalking. It adheres to hot surfaces and can be sprayed in the cold (to 25 degrees Fahrenheit). Once applied, the paint dries in under 5 minutes.