Learn More
Enterprise Technology Solutions Rust-Oleum Marking Paint Spray, Water-Based, 17 oz, APWA Caution BE
SDP
Catalog No. 5000829617
Encompass_Preferred
Industrial Choice Marking Spray Paint features a water-based formula that creates ultra-crisp marks for easy-to-read results. Precision tip sprays accurately onto surfaces with virtually no overspray for consistent application. Bright, durable color resists early fading and chalking to ensure long-lasting visibility. Easy-to-use design dries in under 5 minutes and adheres to hot and cold surfaces. Inverted marking spray paint is perfect for use on concrete, pavement, grass, gravel and more.
Catalog No. 50-008-29617 Supplier Enterprise Technology Solutions Supplier No. RST203031
Industrial Choice Marking Spray Paint features a water-based formula that creates ultra-crisp marks for easy-to-read results. Precision tip sprays accurately onto surfaces with virtually no overspray for consistent application. Bright, durable color resists early fading and chalking to ensure long-lasting visibility. Easy-to-use design dries in under 5 minutes and adheres to hot and cold surfaces. Inverted marking spray paint is perfect for use on concrete, pavement, grass, gravel and more.