Learn More
Enterprise Technology Solutions Rust-Oleum Spray Paint,HighHeat,Rus-Prevent,1200F,Semi-Gloss,12oz,Black
SDP
Catalog No. 5000829088
Encompass_Preferred
High-Heat Spray Paint with rust-preventive enamel delivers deep color and rich sheen to protect indoor and outdoor high-heat applications. Durable, semi-gloss finish retains excellent color retention after repeated heating. High-heat sprays withstand temperatures up to 1,200 degrees Fahrenheit. Use on the exterior of barbecue grills, wood stoves, radiators, fire pits, automotive parts and more. Avoid use in areas directly exposed to flames.
Catalog No. 50-008-29088 Supplier Enterprise Technology Solutions Supplier No. RST241169
High-Heat Spray Paint with rust-preventive enamel delivers deep color and rich sheen to protect indoor and outdoor high-heat applications. Durable, semi-gloss finish retains excellent color retention after repeated heating. High-heat sprays withstand temperatures up to 1,200 degrees Fahrenheit. Use on the exterior of barbecue grills, wood stoves, radiators, fire pits, automotive parts and more. Avoid use in areas directly exposed to flames.