Learn More
Enterprise Technology Solutions Rust-Oleum Spray Paint,Marking,M1600,SolventBased,Precision,17oz,FLOE
SDP
Catalog No. 5000829618
Encompass_Preferred
Industrial Choice M1600 System Precision Line Inverted Marking Spray Paint lets you easily create bold, durable marks for construction, excavation, surveying and more. Solvent-based formula dries quickly and resists early fading and chalking. Marker paint adheres to both hot and cold surfaces with ease. Use on grass, gravel, dirt, concrete and more.
Catalog No. 50-008-29618 Supplier Enterprise Technology Solutions Supplier No. RST203027V
Industrial Choice M1600 System Precision Line Inverted Marking Spray Paint lets you easily create bold, durable marks for construction, excavation, surveying and more. Solvent-based formula dries quickly and resists early fading and chalking. Marker paint adheres to both hot and cold surfaces with ease. Use on grass, gravel, dirt, concrete and more.