Learn More
Ampco Rust Scraper
SDP
Catalog No. 19173528 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
23 -1/2 in.
31 -1/2 in.
47 -1/4 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-173-528 47 -1/4 in.
19-173-525 23 -1/2 in.
19-173-526 31 -1/2 in.
19-173-527 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-173-528 Supplier Ampco Supplier No. 8606
Add to Cart
Add to Cart
Used to remove rust and corrosion from metal surfaces. Non-magnetic and corrosion-resistant.
  • 3 in. blade width.
  • Made in the UK.
  • Comes in 4 different lengths.
Specifications
Product Type Rust Scraper
Length (English) 47 -1/4 in.
Quantity 1 Ea.