Learn More
Chemglass Life Sciences Rushton Style Impeller Blade for 5-7L Bio Reactors, 60mm Dia for 8mm OD Shaft
SDP
Catalog No. 501294457
Encompass
Rushton Style Impeller Blade for 5-7L Bio Reactors, 60mm Dia. The 3 through 7 Liter sizes fit the 8mm Shaft, while the 15 liter fits the 15mm shaft. These blades are used primarily for bacterial cell culture work.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4457 Supplier Chemglass Life Sciences Supplier No. CLS138207
Rushton Style Impeller Blade for 5-7L Bio Reactors, 60mm Dia. The 3 through 7 Liter sizes fit the 8mm Shaft, while the 15 liter fits the 15mm shaft. These blades are used primarily for bacterial cell culture work.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.