Learn More
Medchemexpress LLC HY-N0147 10mg Medchemexpress, Rutaecarpine CAS:84-26-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN014710MG
Encompass_Preferred
Medchemexpress, HY-N0147 10mg Rutaecarpine CAS:84-26-4 Rutaecarpine, an alkaloid of Evodia rutaecarpa, is an inhibitor of COX-2 with an IC 50 value of 0.28 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3426