Learn More
Cayman Chemical Rutaecarpine, Purity: ≥98%, CAS: 84-26-4, 10 mg
SDP
Catalog No. 502994209
Encompass_Preferred
A quinazolinone alkaloid with diverse biological activities; inhibits COX-1 and COX-2 in BMMC cells (IC50s = 8.7 and 0.28 µM, respectively); selective for COX-2 in HEK293 cells (IC50s = >400 and 2.8 µM, for COX-1 and COX-2, respectively); slows cancer cell growth in vitro (GI50s = 8.41-31.6 µM); induces vasodilation (0.1-10 µM) in isolated rat aorta; decreases plasma levels of caffeine in rat through CYP1A2 and CYP2E1 metabolism induction at a dose of 80 mg/kg
Catalog No. 50-299-4209 Supplier Cayman Chemical Supplier No. 2289710 MG
A quinazolinone alkaloid with diverse biological activities; inhibits COX-1 and COX-2 in BMMC cells (IC50s = 8.7 and 0.28 µM, respectively); selective for COX-2 in HEK293 cells (IC50s = >400 and 2.8 µM, for COX-1 and COX-2, respectively); slows cancer cell growth in vitro (GI50s = 8.41-31.6 µM); induces vasodilation (0.1-10 µM) in isolated rat aorta; decreases plasma levels of caffeine in rat through CYP1A2 and CYP2E1 metabolism induction at a dose of 80 mg/kg