Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rutaecarpine 98.0+%, TCI America™
SDP

Catalog No. R01021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01021G 1 g
R0102200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R01021G Supplier TCI America Supplier No. R01021G

Chemical Identifiers

CAS 84-26-4
Molecular Formula C18H13N3O
Molecular Weight (g/mol) 287.32
MDL Number MFCD00210551
InChI Key ACVGWSKVRYFWRP-UHFFFAOYSA-N
Synonym 8,13-Dihydroindolo[2′C,3′C:3,4]pyrido[2,1-b]quinazolin-5(7H)-one
PubChem CID 65752
ChEBI CHEBI:8922
IUPAC Name 3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(10),4,6,8,15,17,19-octaen-14-one
SMILES O=C1N2CCC3=C(NC4=CC=CC=C34)C2=NC2=CC=CC=C12

Specifications

Melting Point 262°C
Color Yellow
Quantity 1 g
Formula Weight 287.32
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Rutaecarpine
RTECSNumber NM3454000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.