Learn More
Strem, An Ascensus Company CAS# 7440-18-8. 100g. Ruthenium, 0.5% on alumina. MFCD00011207
SDP
Catalog No. 5090111990
Encompass
CAS# 7440-18-8. 100g. Ruthenium, 0.5% on alumina. MFCD00011207. Molecular Weight: . Molecular Formula: Ru on alumina. Color/form: 1/8'' x 1/8'' pellets Strem# 44-3880. http://www.strem.com/catalog/v/44-3880/
Catalog No. 50-901-11990 Supplier Strem, An Ascensus Company Supplier No. 443880100G
CAS# 7440-18-8. 100g. Ruthenium, 0.5% on alumina. MFCD00011207. Molecular Weight: . Molecular Formula: Ru on alumina. Color/form: 1/8'' x 1/8'' pellets Strem# 44-3880. http://www.strem.com/catalog/v/44-3880/