Learn More
Medchemexpress LLC Ruthenium(2+) bis(2,2'-bipyridine) mcbpy-O-succinimide ester (PF6)2 | 136724-73-7 | MFCD02683415 | 97.0% | 1014.66 g/mol | C36H29F12N7O4P2Ru | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW1277165MG
Encompass_Preferred
Ru(bpy)2(mcbpy-O-Su-ester)(PF6)2 is a ruthenium-based luminescent activated ester dye for bioconjugation and analytical labeling. It is used to covalently tag proteins and probes, to quench quantum dot fluorescence, and as a sensitive electrogenerated chemiluminescence label in detection assays. Supplied as a brown to red solid, the compound contains an N-hydroxysuccinimide (NHS) ester for amine coupling.
Catalog No. 50-002-83535