Learn More
Strem, An Ascensus Company CAS# 7440-18-8. 25g. Ruthenium, 5% on carbon. MFCD00011207
SDP
Catalog No. 5090111996
Encompass
CAS# 7440-18-8. 25g. Ruthenium, 5% on carbon. MFCD00011207. Molecular Weight: . Molecular Formula: Ru on carbon. Color/form: pwdr. Strem# 44-4050. http://www.strem.com/catalog/v/44-4050/
Catalog No. 50-901-11996 Supplier Strem, An Ascensus Company Supplier No. 44405025G
CAS# 7440-18-8. 25g. Ruthenium, 5% on carbon. MFCD00011207. Molecular Weight: . Molecular Formula: Ru on carbon. Color/form: pwdr. Strem# 44-4050. http://www.strem.com/catalog/v/44-4050/