Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium AA Standard, 1000 ppm Ru in 10% HCl, Ricca Chemical
SDP

Catalog No. ARU1KH100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ARU1KH-100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ARU1KH-100 Supplier Ricca Chemical Company Supplier No. ARU1KH100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified Ruthenium AA standard for atomic absorption analysis.

Chemical Identifiers

CAS 14898-67-0 , 7647-01-0 , 7732-18-5
Molecular Formula Cl3Ru
Molecular Weight (g/mol) 207.42
MDL Number MFCD00149844
InChI Key YBCAZPLXEGKKFM-UHFFFAOYSA-K
IUPAC Name ruthenium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Ru
CAS Min % 87.0, 10.32, 0.21
CAS Max % 91.0, 10.74, 0.22
Equivalence 1 mL = 1 mg Ru
Physical Form Liquid
Color Brown/Red
pH < 1
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) Atomic Absorption Spectrophotometer
Solubility Miscible with water
Solution Type Calibration Standard
Type Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Ruthenium AA Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.