Learn More
ALADDIN SCIENTIFIC CORPORATION Ruthenium acetylacetonate, 1g
Catalog No. 5000082533
Encompass_Preferred
Catalyzes the hydrogenation of dimethyl oxalate to ethylene glycol under mild conditions Product classM-O Homogeneous Catalysts Acetylacetonate LigandsChemical propertiesChemical formulaC15H21O6RuEmpirical formula[Ru(acac)3]Molecular weight398 40MetalRuTheoretical metal content25Physical statepowderColorred
Catalog No. 50-000-82533 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. R1185021G
Catalyzes the hydrogenation of dimethyl oxalate to ethylene glycol under mild conditions Product classM-O Homogeneous Catalysts Acetylacetonate LigandsChemical propertiesChemical formulaC15H21O6RuEmpirical formula[Ru(acac)3]Molecular weight398 40MetalRuTheoretical metal content25Physical statepowderColorred