Learn More
Sigma Aldrich Ruthenium On Alumina
Catalog No. 206199250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
50 g
250 g
Catalog No. 20-619-9250G
Supplier: Sigma Aldrich 206199250G
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Ruthenium on alumina (Ru/Al2O3) (0.5 wt. % loading, pellets, 3.2 mm) can be used as a catalyst for the production of methane via hydrogenation of carbon dioxide. It can also be used as a catalyst in the reduction of CO2.
Specifications
MDL Number MFCD00011207
Quantity 250 g
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.