Learn More
Sigma Aldrich Ruthenium on alumina
Catalog No. 43991625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
25 g
Catalog No. Quantity
43-991-625G 25 g
1 options
Catalog No. 43-991-625G Supplier Sigma Aldrich Supplier No. 43991625G
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Ruthenium on alumina (Ru/Al2O3) (5 wt. % loading, powder, reduced, dry) can be used as a catalyst for the production of methane via hydrogenation of carbon dioxide. It can also be used as a catalyst in the reduction of CO2.
Specifications
MDL Number MFCD00011207
Quantity 25 g