Learn More
Strem, An Ascensus Company CAS 1295649-40-9. 1g. Carbonylchlorohydrido[bis(2-(diphenylphosphinoethyl)amino]ruthenium(II) min.98% Ru-MACHO. MFCD22377801
SDP
Supplier:  Strem, An Ascensus Company 4400711G
Encompass
CAS 1295649-40-9. 1g. Carbonylchlorohydrido[bis(2-(diphenylphosphinoethyl)amino]ruthenium(II) min.98% Ru-MACHO. MFCD22377801. Molecular Weight 607.03. Molecular Formula C29H30ClNOP2Ru. Color/form white to yellow pwdr. Strem 44-0071. http//www.strem.com/catalog/v/44-0071/
Catalog No. 50-211-0816