Learn More
Strem, An Ascensus Company CAS . 500mg. Dichloro[(S)-(-)-22-bis(diphenylphosphino)-11-binaphthyl][(S)-(-)-2-(-(t-butyl)methanamine)-1H-benzimidazole]ruthenium(II) min. 95%. MFCD16038122
SDP
Catalog No. 502110828
Encompass
500mg. Dichloro[(S)-(-)-22-bis(diphenylphosphino)-11-binaphthyl][(S)-(-)-2-(-(t-butyl)methanamine)-1H-benzimidazole]ruthenium(II) min. 95%. MFCD16038122. Molecular Weight 997.93. Molecular Formula C56H49Cl2N3P2Ru. Color/form yellow-brown pwdr. Strem 44-0925. http//www.strem.com/catalog/v/44-0925/
Catalog No. 50-211-0828 Supplier Strem, An Ascensus Company Supplier No. 440925500MG
500mg. Dichloro[(S)-(-)-22-bis(diphenylphosphino)-11-binaphthyl][(S)-(-)-2-(-(t-butyl)methanamine)-1H-benzimidazole]ruthenium(II) min. 95%. MFCD16038122. Molecular Weight 997.93. Molecular Formula C56H49Cl2N3P2Ru. Color/form yellow-brown pwdr. Strem 44-0925. http//www.strem.com/catalog/v/44-0925/