Learn More
Strem, An Ascensus Company CAS 1190427-50-9. 500mg. Tricyclohexylphosphine[45-dimethyl-13-bis(246-trimethylphenyl)imidazol-2-ylidene][2-thienylmethylene]ruthenium(II) dichloride min. 95% [catMETium RF 3]. MFCD16038135
SDP
Catalog No. 502110836
Encompass
CAS 1190427-50-9. 500mg. Tricyclohexylphosphine[45-dimethyl-13-bis(246-trimethylphenyl)imidazol-2-ylidene][2-thienylmethylene]ruthenium(II) dichloride min. 95% [catMETium RF 3]. MFCD16038135. Molecular Weight 881.04. Molecular Formula C46H65Cl2N2PRuS. Color/form violet to brown pwdr. Strem 44-7790. http//www.strem.com/catalog/v/44-7790/
Catalog No. 50-211-0836 Supplier Strem, An Ascensus Company Supplier No. 447790500MG
CAS 1190427-50-9. 500mg. Tricyclohexylphosphine[45-dimethyl-13-bis(246-trimethylphenyl)imidazol-2-ylidene][2-thienylmethylene]ruthenium(II) dichloride min. 95% [catMETium RF 3]. MFCD16038135. Molecular Weight 881.04. Molecular Formula C46H65Cl2N2PRuS. Color/form violet to brown pwdr. Strem 44-7790. http//www.strem.com/catalog/v/44-7790/