Learn More
Strem, An Ascensus Company CAS# 12289-94-0. 5G. Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II), min. 97%. MFCD00216965
SDP
Catalog No. 501963533
Encompass
CAS# 12289-94-0. 5G. Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II), min. 97%. MFCD00216965. Molecular Formula: (C4H7)2(C8H12)Ru. Molecular Weight: 319.45. Color/Form: tan to brown pwdr. Stability: store cold (-18°C). Strem# 44-0046. www.strem.com/catalog/v3/44-0046/
Catalog No. 50-196-3533 Supplier Strem, An Ascensus Company Supplier No. 4400465G
CAS# 12289-94-0. 5G. Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II), min. 97%. MFCD00216965. Molecular Formula: (C4H7)2(C8H12)Ru. Molecular Weight: 319.45. Color/Form: tan to brown pwdr. Stability: store cold (-18°C). Strem# 44-0046. www.strem.com/catalog/v3/44-0046/