Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(II) Nitrosyl Chloride Monohydrate 97.0+%, TCI America™
SDP

Catalog No. R00861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R00861G Supplier TCI America Supplier No. R00861G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18902-42-6
Molecular Formula Cl3NORu
Molecular Weight (g/mol) 237.43
MDL Number MFCD00150427
InChI Key AUWSZCCJRSCQEP-UHFFFAOYSA-K
Synonym Trichloronitrosylruthenium(II)

Specifications

Color Red-Yellow
Physical Form Crystalline Powder
Quantity 1 g
Formula Weight 237.43
Percent Purity ≥97.0% (T)
Chemical Name or Material Ruthenium(II) Nitrosyl Chloride Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.