Learn More
Medchemexpress LLC Ruthenium(III) acetylacetonate | 14284-93-6 | MFCD00000030 | 99.9% | 398.39 | C15H21O6Ru | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0209581G
Encompass_Preferred
Ru(acac)3 (tris(acetylacetonato)ruthenium(III), CAS 14284-93-6) is a coordination compound used as a biochemical reagent and catalyst in materials and life-science research. Supplied for research use only, the compound is reported at high purity and has defined storage recommendations to preserve stability.
  • High purity (99.9%) suitable for analytical and synthetic applications.
  • Stable at room temperature when protected from light and stored under nitrogen.
  • Solvent stability: -80°C for up to 2 years, -20°C for up to 1 year.
  • Useful as a catalyst and precursor for ruthenium-containing materials.
  • Provided in small laboratory pack sizes for research workflows.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-003-01392