Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ruthenium(III) Chloride

Catalog No. 20852310G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-852-310G 10 g
20-852-350G 50 g
20-852-32G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 20-852-310G Supplier Sigma Aldrich Supplier No. 20852310G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst used in the oxidative cyclization of 1,7-dienes to oxepane diols. Hydroxylates tertiary carbon-hydrogen bonds of cyclic ethers with periodate or bromate.insoluble form

Specifications

CAS 10049-08-8
Density 3.11 g/mL at 25°C
Molecular Formula Cl3Ru
Linear Formula RuCl3
MDL Number MFCD00011208
Quantity 10 g
Molecular Weight (g/mol) 207.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.