Learn More
Strem, An Ascensus Company CAS# 10049-08-8. Ruthenium(III) chloride, anhydrous MFCD00011208. 1G
SDP
Catalog No. 5090112009
Encompass
CAS# 10049-08-8. 1G. Ruthenium(III) chloride, anhydrous. MFCD00011208. Molecular Formula: Cl3Ru Molecular Weight: 207.43. Color/Form: black pwdr. Stability: hygroscopic Strem#: 44-5850 www.strem.com/catalog/v3/44-5850/
Catalog No. 50-901-12009 Supplier Strem, An Ascensus Company Supplier No. 4458501G
CAS# 10049-08-8. 1G. Ruthenium(III) chloride, anhydrous. MFCD00011208. Molecular Formula: Cl3Ru Molecular Weight: 207.43. Color/Form: black pwdr. Stability: hygroscopic Strem#: 44-5850 www.strem.com/catalog/v3/44-5850/