Learn More
Strem, An Ascensus Company CAS# 14898-67-0. 1g. Ruthenium(III) chloride hydrate (40-43% Ru) (99.9%-Ru). MFCD00149844
SDP
Supplier:  Strem, An Ascensus Company 4458801G
Encompass
CAS# 14898-67-0. 1g. Ruthenium(III) chloride hydrate (40-43% Ru) (99.9%-Ru). MFCD00149844. Molecular Weight: 207.43. Molecular Formula: RuCl3∙XH2O. Color/form: black pwdr. Strem# 44-5880. http://www.strem.com/catalog/v/44-5880/
Catalog No. 50-901-12011