Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(III) nitrosylsulfate, Premion™, 99.99% (metals basis)

Catalog No. AA4343606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4343606 5 g
AA4343603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4343606 Supplier Thermo Scientific Chemicals Supplier No. 04343606
Add to Cart

missing translation for 'validMainframeMsg'

Electroplating, coating | [Ru(NO)]2(SO4)3

Ruthenium(III) nitrosylsulfate is used in Electroplating, coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula [Ru(NO)]2(SO4)3
MDL Number MFCD02684508

Specifications

Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water
Formula Weight 550.34
Percent Purity 99.99%
Grade Premion
Assay (metals basis)
Chemical Name or Material Ruthenium (III) nitrosylsulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.