Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ruthenium(IV) oxide, Electronic Grade, Premion™, 99.95% (metals basis), Ru typically 74%

Catalog No. AA4033609
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4033609 10 g
AA4033604 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4033609 Supplier Thermo Scientific Chemicals Supplier No. 04033609
Add to Cart

missing translation for 'validMainframeMsg'

For Semiconductor Industry Use For the Manufacture of Resistor Pastes | CAS: 12036-10-1 | O2Ru | 133.07 g/mol

The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. It is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-10-1
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00011210
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
Synonym ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
PubChem CID 82848
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O

Specifications

Melting Point 1200° (sublimation)
Color Black
Physical Form Sub-micron Powder
Quantity 10 g
Odor Odorless
Sensitivity Hygroscopic
Appearance Black powder
Assay Percent Range 99.95% (metals basis)
Density 6.97
Solubility Information Insoluble in water,acids. Soluble in fused alkalis.
Formula Weight 133.07
Grade Electronic
Chemical Name or Material Ruthenium(IV) oxide
Show More Show Less
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 234-840-6
RTECSNumber VM2654000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.