Learn More
Sigma Aldrich Ruthenium On Alumina
Catalog No. 20619950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-619-950G 50 g
20-619-9250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-619-950G Supplier Sigma Aldrich Supplier No. 20619950G
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Ruthenium on alumina (Ru/Al2O3) (0.5 wt. % loading, pellets, 3.2 mm) can be used as a catalyst for the production of methane via hydrogenation of carbon dioxide. It can also be used as a catalyst in the reduction of CO2.
Specifications
MDL Number MFCD00011207
Quantity 50 g