Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ruthenium On Carbon

Catalog No. 90804510G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-804-510G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-804-510G Supplier Sigma Aldrich Supplier No. 90804510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Evonik Noblyst™ P3059 5% Ru is a ruthenium on carbon catalyst. It has been shown to be effective in the following reactions:hydrogenation of aldehydes and ketoneshydrogenation of sugarshydrogenation of carboxylic acids, esters, and anhydridespartial hydrogenation of aromaticsUnit weight is of net dry catalyst.Noblyst is a registered trademark of Evonik Degussa GmbH

Specifications

CAS 2300178-66-7
Melting Point >300°C
Molecular Formula C12H12Cl2N2Ni -+ xH2O
Quantity 500 mg
Synonym (4,4'-dmbpy)NiCl2
Molecular Weight (g/mol) 313.84 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.