Learn More
Advanced Biomatrix Ruthenium Photoinitiator Kit, non-sterile
Supplier:  Advanced Biomatrix 52481KIT
Encompass_Preferred
Ruthenium is a water soluble photoinitiator that utilizes visible light (400-450nm) to covalently crosslink free tyrosine and acryl groups. Ruthenium has been tested on collagen type I, gelatin, silk fibroin, methacrylated hyaluronic acid, methacrylated gelatin, methacrylated collagen type I and PEGDA.
Catalog No. 50-225-5919