Learn More
Strem, An Ascensus Company CAS# 7440-18-8. 1g. Ruthenium powder (99.99%). MFCD00011207
SDP
Catalog No. 5090112007
Encompass
CAS# 7440-18-8. 1g. Ruthenium powder (99.99%). MFCD00011207. Molecular Weight: 101.07. Molecular Formula: Ru. Color/form: -200 mesh pwdr. Strem# 44-4350. http://www.strem.com/catalog/v/44-4350/
Catalog No. 50-901-12007 Supplier Strem, An Ascensus Company Supplier No. 4443501G
CAS# 7440-18-8. 1g. Ruthenium powder (99.99%). MFCD00011207. Molecular Weight: 101.07. Molecular Formula: Ru. Color/form: -200 mesh pwdr. Strem# 44-4350. http://www.strem.com/catalog/v/44-4350/