Learn More
Apexbio Technology LLC Ruthenium Red 100mg
SDP
Catalog No. 502240134
Encompass_Preferred
Ruthenium Red (CAS 11103-72-3) is a potent inhibitor of calcium (Ca ) transport across biological membranes including mitochondrial erythrocyte and rabbit skeletal muscle sarcoplasmic reticulum membranes It acts by blocking Ca uptake at specific binding sites one high-affinity site (K sub m /sub 4 5 M) and one lower-affinity site (K sub m /sub 2 0 mM) associated with the Ca -ATPase transmembrane domain Incubation of sarcoplasmic reticulum vesicles with micromolar concentrations of Ruthenium Red significantly reduces Ca binding capacity Additionally Ruthenium Red suppresses capsaicin-induced plasma extravasation in rat trachea in a dose-dependent manner This compound is widely used to investigate Ca signaling transport mechanisms and neurogenic inflammation in biomedical research
This item is not returnable. View return policy
Catalog No. 50-224-0134 Supplier Apexbio Technology LLC Supplier No. B6740100
Add to Cart
Add to Cart
Ruthenium Red (CAS 11103-72-3) is a potent inhibitor of calcium (Ca ) transport across biological membranes including mitochondrial erythrocyte and rabbit skeletal muscle sarcoplasmic reticulum membranes It acts by blocking Ca uptake at specific binding sites one high-affinity site (K sub m /sub 4 5 M) and one lower-affinity site (K sub m /sub 2 0 mM) associated with the Ca -ATPase transmembrane domain Incubation of sarcoplasmic reticulum vesicles with micromolar concentrations of Ruthenium Red significantly reduces Ca binding capacity Additionally Ruthenium Red suppresses capsaicin-induced plasma extravasation in rat trachea in a dose-dependent manner This compound is widely used to investigate Ca signaling transport mechanisms and neurogenic inflammation in biomedical research