Learn More
Cayman Chemical Ruthenium Red 100mg
SDP
Catalog No. 502953858
Encompass_Preferred
A polycationic dye that inhibits sarcoplasmic reticulum Ca2+ release, blocks Ca2+ uptake and release from mitochondria, and prevents Ca2+ release from ryanodine-sensitive intracellular stores in skeletal and cardiac muscle (Kis = 16 nm - 5.9 μM, depending on Ca2+ concentration)
Catalog No. 50-295-3858 Supplier Cayman Chemical Supplier No. 14339100 MG
A polycationic dye that inhibits sarcoplasmic reticulum Ca2+ release, blocks Ca2+ uptake and release from mitochondria, and prevents Ca2+ release from ryanodine-sensitive intracellular stores in skeletal and cardiac muscle (Kis = 16 nm - 5.9 μM, depending on Ca2+ concentration)