Learn More
Enzo Life Sciences Ruthenium red (1g). CAS: 11103-72-3
SDP
Catalog No. 502009527
Encompass_Preferred
Blocks TRPV1. Capsaicin and calcium antagonist. Inhibitor of Ca2+/Mg2+-ATPase. Purity: ≥90% (metals basis). Appearance: Dark red to purple crystalline solid. Solubility: Soluble in water (10mg/ml) or ammonia. Long Term Storage: Ambient.
Catalog No. 50-200-9527 Supplier Enzo Life Sciences Supplier No. ALX550295G001
Blocks TRPV1. Capsaicin and calcium antagonist. Inhibitor of Ca2+/Mg2+-ATPase. Purity: ≥90% (metals basis). Appearance: Dark red to purple crystalline solid. Solubility: Soluble in water (10mg/ml) or ammonia. Long Term Storage: Ambient.