Learn More
TARGETMOL CHEMICALS INC Ruthenium Red 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T23280500MG
Encompass_Preferred
Also available in 100 mg and bulk. Please contact Fisher for quotes. Ruthenium Red (Ammoniated ruthenium oxychloride) is a polycationic dye widely used for electron microscopy (EM) of cells, tissues and vegetative bacteria. Ruthenium red strongly reacts with phospholipids and fatty acids and binds to acidic mucopolysaccharides. Ruthenium red is a L-type calcium current (ICa) blocker, blocks Ca2+ uptake and release, and voltage-sensitive Ca2+ channels Purity 95%
Catalog No. 50-287-7546